Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271472.1 | NP_001258401.1 | 14 - 15 | 1768 | 77 | |
NM_001271473.1 | NP_001258402.1 | 11 - 12 | 1435 | 77 | ||
NM_022723.3 | NP_073560.2 | 13 - 14 | 1678 | 77 | ||
GenBank mRNA | AF276425.1 | - | 13 - 14 | 1545 | 77 | |
AK041590.1 | - | 13 - 14 | 1661 | 77 | ||
BC080278.1 | - | 11 - 12 | 1403 | 30354974 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |