Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205345.1 | NP_001192274.1 | 14 - 15 | 1939 | 91 | |
NM_001205346.1 | NP_001192275.1 | 14 - 15 | 1933 | 91 | ||
NM_194342.3 | NP_919323.2 | 13 - 14 | 1843 | 91 | ||
GenBank mRNA | AK047688.1 | - | 2 - 3 | 286 | 91 | |
AK128958.1 | - | 13 - 14 | 1837 | 91 | ||
AK156246.1 | - | 14 - 15 | 1940 | 91 | ||
AK171058.1 | - | 14 - 15 | 1902 | 91 | ||
AK172398.1 | - | 14 - 15 | 1941 | 91 | ||
AK172996.1 | - | 12 - 13 | 2543 | 91 | ||
AY682987.1 | - | 13 - 14 | 1699 | 91 | ||
BC023965.1 | - | 3 - 4 | 491 | 91 | ||
BC098208.1 | - | 13 - 14 | 2184 | 6827666 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |