Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207225.1 | NP_997108.1 | 13 - 14 | 2066 | 85 | |
GenBank mRNA | AK029933.1 | - | 10 - 11 | 1603 | 85 | |
AK155250.1 | - | 14 - 15 | 2252 | 85 | ||
BC066052.1 | - | 13 - 14 | 2066 | 6827645 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |