Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134252.3 | NP_599013.1 | 13 - 14 | 1832 | 78 | |
GenBank mRNA | AF481480.2 | - | 2101 | 78 | ||
AY095352.1 | - | 11 - 12 | 1666 | 78 | ||
BC117934.1 | - | 13 - 14 | 1832 | 40097778 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |