Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025454.2 | NP_079730.1 | 3 - 4 | 313 | 90 | |
GenBank mRNA | AK006421.1 | - | 3 - 4 | 412 | 90 | |
AK007536.1 | - | 3 - 4 | 304 | 90 | ||
AK009312.1 | - | 3 - 4 | 297 | 90 | ||
AK028010.1 | - | 3 - 4 | 291 | 90 | ||
AK028397.1 | - | 3 - 4 | 314 | 90 | ||
AK145695.1 | - | 3 - 4 | 286 | 90 | ||
BC064674.1 | - | 3 - 4 | 312 | 6400183 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |