Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031621.2 | NP_001026792.2 | 15 - 16 | 2386 | 116 | |
GenBank mRNA | AB112584.1 | - | 15 - 16 | 2386 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |