Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700014367.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014367.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014367.mm.1 |
TC1700001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014371.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014371.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014371.mm.1 |
TC1700001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014368.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014368.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014368.mm.1 |
TC1700001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014369.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014369.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014369.mm.1 |
TC1700001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014370.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014370.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014370.mm.1 |
TC1700001624.mm.1 |
MTA TC |