Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009324.2 | NP_033350.2 | 1 - 2 | 887 | 66 | |
GenBank mRNA | AK137225.1 | - | 1 - 2 | 93 | 66 | |
AK149348.1 | - | 1 - 2 | 695 | 66 | ||
U15566.1 | - | 1 - 2 | 423 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |