Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144810.5 | NP_659059.1 | 2 - 3 | 1193 | 79 | |
GenBank mRNA | AK043039.1 | - | 2 - 3 | 531 | 79 | |
AK044318.1 | - | 2 - 3 | 769 | 79 | ||
AK080356.1 | - | 2 - 3 | 1193 | 79 | ||
BC011135.1 | - | 2 - 3 | 758 | 4191503 | 79 | |
BC023173.1 | - | 2 - 3 | 392 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |