Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021530.2 | NP_067505.2 | 17 - 18 | 2467 | 93 | |
GenBank mRNA | AF224508.1 | - | 17 - 18 | 2413 | 93 | |
AK034237.1 | - | 17 - 18 | 2469 | 93 | ||
AK039404.1 | - | 17 - 18 | 2432 | 93 | ||
AK047863.1 | - | 10 - 11 | 1401 | 93 | ||
AK171610.1 | - | 16 - 17 | 2243 | 93 | ||
AK173014.1 | - | 17 - 18 | 2301 | 93 | ||
AK183206.1 | - | 2 - 3 | 213 | 93 | ||
BC030388.1 | - | 17 - 18 | 2429 | 5401143 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |