Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021530.2 | NP_067505.2 | 6 - 7 | 946 | 61 | |
GenBank mRNA | AK034237.1 | - | 6 - 7 | 948 | 61 | |
AK039404.1 | - | 6 - 7 | 911 | 61 | ||
AK171610.1 | - | 5 - 6 | 722 | 61 | ||
AK173014.1 | - | 6 - 7 | 780 | 61 | ||
BC030388.1 | - | 6 - 7 | 908 | 5401143 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |