Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E4f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF126967.1 | - | 11 - 12 | 1804 | 71 | |
AK033285.1 | - | 11 - 12 | 1813 | 71 | ||
AK077014.1 | - | 72 | ||||
AK144176.1 | - | 11 - 12 | 1761 | 71 | ||
BC005634.1 | - | 11 - 12 | 1735 | 71 | ||
BC031757.1 | - | 10 - 11 | 1945 | 71 | ||
BC046459.1 | - | 11 - 12 | 1874 | 5322519 | 71 | |
BC057011.1 | - | 10 - 11 | 1467 | 71 | ||
BC071228.1 | - | 11 - 12 | 1791 | 4915902 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |