Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700014465.mm.1 |
NM_001252463.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014465.mm.1 |
NM_001252463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014465.mm.1 |
NM_001252464.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014465.mm.1 |
NM_001252464.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014465.mm.1 |
NM_001252465.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014465.mm.1 |
NM_001252465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014465.mm.1 |
NM_019988.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014465.mm.1 |
NM_019988.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014465.mm.1 |
TC1700001631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014467.mm.1 |
NM_001252463.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014467.mm.1 |
NM_001252463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014467.mm.1 |
NM_001252464.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014467.mm.1 |
NM_001252464.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014467.mm.1 |
NM_001252465.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014467.mm.1 |
NM_001252465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014467.mm.1 |
NM_019988.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014467.mm.1 |
NM_019988.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014467.mm.1 |
TC1700001631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014461.mm.1 |
NM_001252463.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014461.mm.1 |
NM_001252463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014461.mm.1 |
NM_001252464.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014461.mm.1 |
NM_001252464.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014461.mm.1 |
NM_019988.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014461.mm.1 |
NM_019988.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014461.mm.1 |
TC1700001631.mm.1 |
MTA TC |