Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080773.2 | NP_001074242.1 | 2 - 3 | 431 | 148 | |
NM_001286662.1 | NP_001273591.1 | 2 - 3 | 349 | 148 | ||
NM_011062.4 | NP_035192.2 | 2 - 3 | 431 | 148 | ||
GenBank mRNA | AF079535.1 | - | 2 - 3 | 388 | 148 | |
AF126294.1 | - | 2 - 3 | 372 | 148 | ||
AK147149.1 | - | 2 - 3 | 423 | 148 | ||
AK147835.1 | - | 2 - 3 | 406 | 148 | ||
AK149162.1 | - | 2 - 3 | 364 | 148 | ||
AK149289.1 | - | 2 - 3 | 389 | 148 | ||
AK162677.1 | - | 2 - 3 | 391 | 148 | ||
AK165564.1 | - | 2 - 3 | 368 | 148 | ||
BC146001.1 | - | 2 - 3 | 368 | 40131304 | 148 | |
BC146003.1 | - | 2 - 3 | 368 | 40131306 | 148 | |
CJ046953.1 | - | 2 - 3 | 350 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |