Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eci1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010023.4 | NP_034153.2 | 5 - 6 | 545 | 59 | |
GenBank mRNA | AK004838.1 | - | 5 - 6 | 546 | 59 | |
AK029481.1 | - | 5 - 6 | 546 | 59 | ||
AK077386.1 | - | 5 - 6 | 536 | 59 | ||
BC022712.1 | - | 5 - 6 | 529 | 4487371 | 59 | |
BC054444.1 | - | 5 - 6 | 532 | 6476161 | 59 | |
Z14049.1 | - | 5 - 6 | 525 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |