Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039363.2 | NP_001034452.1 | 26 - 27 | 3120 | 71 | |
NM_001286713.1 | NP_001273642.1 | 27 - 28 | 3249 | 71 | ||
NM_001286714.1 | NP_001273643.1 | 27 - 28 | 3244 | 71 | ||
NM_001286716.1 | NP_001273645.1 | 26 - 27 | 3115 | 71 | ||
NM_001286718.1 | NP_001273647.1 | 26 - 27 | 3117 | 71 | ||
NM_001286720.1 | NP_001273649.1 | 27 - 28 | 3249 | 71 | ||
NM_011647.3 | NP_035777.2 | 27 - 28 | 3249 | 71 | ||
GenBank mRNA | AF132986.1 | - | 23 - 24 | 2777 | 71 | |
AK143799.1 | - | 26 - 27 | 3118 | 71 | ||
AK147302.1 | - | 27 - 28 | 3190 | 71 | ||
AK147487.1 | - | 27 - 28 | 3218 | 71 | ||
AK147874.1 | - | 26 - 27 | 3084 | 71 | ||
AK147908.1 | - | 27 - 28 | 3250 | 71 | ||
AK148062.1 | - | 27 - 28 | 3241 | 71 | ||
AK148072.1 | - | 26 - 27 | 3017 | 71 | ||
AK170589.1 | - | 26 - 27 | 3111 | 71 | ||
AK171007.1 | - | 26 - 27 | 3086 | 71 | ||
AK216627.1 | - | 1 - 2 | 169 | 71 | ||
AK217739.1 | - | 1 - 2 | 185 | 71 | ||
BC052207.1 | - | 13 - 14 | 1568 | 71 | ||
BC052449.1 | - | 26 - 27 | 3096 | 6409037 | 71 | |
BC060701.1 | - | 26 - 27 | 3096 | 6409037 | 71 | |
U37775.1 | - | 26 - 27 | 3161 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |