Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027008.2 | NP_081284.2 | 1 - 2 | 309 | 71 | |
GenBank mRNA | AK011637.1 | - | 1 - 2 | 44 | 71 | |
AK052960.1 | - | 1 - 2 | 311 | 71 | ||
AK129077.1 | - | 1 - 2 | 269 | 71 | ||
AK143644.1 | - | 1 - 2 | 277 | 71 | ||
BC021777.1 | - | 1 - 2 | 261 | 4215964 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |