Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040187.1 | NP_001035277.1 | 4 - 5 | 935 | 79 | |
NM_199079.1 | NP_951061.1 | 4 - 5 | 754 | 79 | ||
NM_199080.2 | NP_951062.1 | 4 - 5 | 935 | 79 | ||
GenBank mRNA | AK039888.1 | - | 4 - 5 | 754 | 79 | |
AK151061.1 | - | 4 - 5 | 710 | 79 | ||
AK152749.1 | - | 4 - 5 | 710 | 79 | ||
AK152831.1 | - | 4 - 5 | 710 | 79 | ||
AK161001.1 | - | 4 - 5 | 751 | 79 | ||
AK161012.1 | - | 4 - 5 | 750 | 79 | ||
BC062910.1 | - | 4 - 5 | 740 | 6816663 | 79 | |
BC096036.1 | - | 4 - 5 | 935 | 30549388 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |