Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145139.2 | NP_660121.2 | 5 - 6 | 471 | 155 | |
GenBank mRNA | AJ310346.1 | - | 5 - 6 | 481 | 155 | |
AK032936.1 | - | 5 - 6 | 459 | 155 | ||
AK075584.1 | - | 5 - 6 | 477 | 155 | ||
AK150895.1 | - | 5 - 6 | 466 | 155 | ||
AK151175.1 | - | 5 - 6 | 466 | 155 | ||
AK167640.1 | - | 5 - 6 | 155 | |||
AK168009.1 | - | 5 - 6 | 469 | 155 | ||
AK168295.1 | - | 5 - 6 | 468 | 155 | ||
BC024463.1 | - | 5 - 6 | 471 | 4975621 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |