Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145139.2 | NP_660121.2 | 6 - 7 | 547 | 144 | |
GenBank mRNA | AA038997.1 | - | 2 - 3 | 146 | 144 | |
AJ310346.1 | - | 6 - 7 | 557 | 144 | ||
AK032936.1 | - | 6 - 7 | 535 | 144 | ||
AK075584.1 | - | 6 - 7 | 553 | 144 | ||
AK150895.1 | - | 6 - 7 | 542 | 144 | ||
AK151175.1 | - | 6 - 7 | 542 | 144 | ||
AK167640.1 | - | 6 - 7 | 542 | 144 | ||
AK168009.1 | - | 6 - 7 | 545 | 144 | ||
AK168295.1 | - | 6 - 7 | 544 | 144 | ||
BC024463.1 | - | 6 - 7 | 547 | 4975621 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |