Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110891.1 | 2 - 3 | 386 | 96 | ||
GenBank mRNA | BB635805.1 | - | 2 - 3 | 388 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |