Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010668.2 | NP_034798.2 | 6 - 7 | 1379 | 90 | |
GenBank mRNA | AK132476.1 | - | 6 - 7 | 1381 | 90 | |
AK161078.1 | - | 6 - 7 | 1378 | 90 | ||
AK161098.1 | - | 6 - 7 | 1381 | 90 | ||
BC120485.1 | - | 6 - 7 | 1345 | 40129408 | 90 | |
BC139485.1 | - | 6 - 7 | 1345 | 8860587 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |