Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021279.4 | NP_067254.1 | 2 - 3 | 689 | 94 | |
GenBank mRNA | AK141669.1 | - | 2 - 3 | 544 | 94 | |
BC005449.1 | - | 2 - 3 | 678 | 3493095 | 94 | |
M11943.1 | - | 2 - 3 | 541 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |