Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Troap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162506.1 | NP_001155978.1 | 4 - 5 | 595 | 74 | |
NM_030159.1 | NP_084435.1 | 4 - 5 | 620 | 74 | ||
GenBank mRNA | AK050381.1 | - | 4 - 5 | 616 | 74 | |
AK077509.1 | - | 4 - 5 | 585 | 74 | ||
BC080746.1 | - | 4 - 5 | 580 | 74 | ||
BC100405.1 | - | 4 - 5 | 549 | 74 | ||
BC145227.1 | - | 4 - 5 | 993 | 9053779 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |