Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C230037L18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK082325.1 | - | 2 - 3 | 1317 | 86 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | arylsulfatase A | Arsa |
AS-A, ASA, AW212749, As-2, As2, TISP73 |
-118.0 | Anti-sense | Chr. 15 - 89472476 - 89477424 on NCBI Build GRCm38 |
Downstream | SH3/ankyrin domain gene 3 | Shank3 |
AI841104, SHANK3c-3, SHANK3c-4, Shank3b |
21.7kb | Sense | Chr. 15 - 89499707 - 89560261 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |