Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700004250.mm.1 |
NM_001081315.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004250.mm.1 |
NM_001081315.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004250.mm.1 |
TC1700000488.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700004246.mm.1 |
NM_001081315.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004246.mm.1 |
NM_001081315.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004246.mm.1 |
TC1700000488.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700004252.mm.1 |
NM_001081315.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004252.mm.1 |
NM_001081315.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004252.mm.1 |
TC1700000488.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700004249.mm.1 |
NM_001081315.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004249.mm.1 |
NM_001081315.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004249.mm.1 |
TC1700000488.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700004253.mm.1 |
NM_001081315.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004253.mm.1 |
NM_001081315.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004253.mm.1 |
TC1700000488.mm.1 |
MTA TC |