Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018857.1 | NP_061345.1 | 6 - 7 | 623 | 64 | |
GenBank mRNA | AK144391.1 | - | 7 - 8 | 555 | 64 | |
AK146799.1 | - | 6 - 7 | 673 | 64 | ||
BC023753.1 | - | 6 - 7 | 627 | 5354484 | 64 | |
D86370.1 | - | 6 - 7 | 623 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |