Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haghl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271433.1 | NP_001258362.1 | 5 - 6 | 640 | 62 | |
NM_001271434.1 | NP_001258363.1 | 5 - 6 | 640 | 62 | ||
NM_001271435.1 | NP_001258364.1 | 5 - 6 | 640 | 62 | ||
NM_001271436.1 | NP_001258365.1 | 4 - 5 | 454 | 62 | ||
NM_001271437.1 | NP_001258366.1 | 4 - 5 | 449 | 62 | ||
NM_026897.3 | NP_081173.1 | 4 - 5 | 625 | 62 | ||
GenBank mRNA | AK005274.1 | - | 4 - 5 | 621 | 62 | |
AK012748.1 | - | 5 - 6 | 637 | 62 | ||
AK021220.1 | - | 4 - 5 | 451 | 62 | ||
AK034798.1 | - | 5 - 6 | 615 | 62 | ||
AK047517.1 | - | 5 - 6 | 595 | 62 | ||
AK154248.1 | - | 4 - 5 | 430 | 62 | ||
BC005586.1 | - | 5 - 6 | 586 | 62 | ||
BC030466.1 | - | 5 - 6 | 625 | 5253213 | 62 | |
BC083322.1 | - | 5 - 6 | 579 | 6820885 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |