Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erbb2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005868.2 | NP_001005868.1 | 13 - 14 | 1534 | 105 | |
NM_001289473.1 | NP_001276402.1 | 13 - 14 | 1534 | 105 | ||
NM_001289474.1 | NP_001276403.1 | 13 - 14 | 1534 | 105 | ||
NM_001289475.1 | NP_001276404.1 | 13 - 14 | 1534 | 105 | ||
GenBank mRNA | AK029054.1 | - | 2 - 3 | 216 | 105 | |
AK122473.1 | - | 13 - 14 | 1213 | 105 | ||
AK169529.1 | - | 13 - 14 | 1463 | 105 | ||
BC141178.1 | - | 13 - 14 | 1243 | 9055790 | 105 | |
BC141188.1 | - | 13 - 14 | 1243 | 9055800 | 105 | |
BC145498.1 | - | 13 - 14 | 1243 | 9054061 | 105 | |
BC145509.1 | - | 13 - 14 | 1243 | 9054072 | 105 | |
EU233994.1 | - | 15 - 16 | 2291 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |