Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009604.3 | NP_033734.3 | 3 - 4 | 255 | 57 | |
GenBank mRNA | AK047974.1 | - | 3 - 4 | 256 | 57 | |
JF520813.1 | - | 3 - 4 | 256 | 57 | ||
JN164665.1 | - | 423 | 57 | |||
JN164666.1 | - | 2 - 3 | 314 | 57 | ||
M30514.1 | - | 3 - 4 | 296 | 57 | ||
X03818.1 | - | 3 - 4 | 252 | 57 | ||
X03819.1 | - | 2 - 3 | 187 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Mus musculus Probe spans exons |