Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28035861 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28035860 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2007384 |
AF019564.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2007384 |
AK009015.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2007384 |
AK075915.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2007384 |
BC060287.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2007384 |
NM_001136080.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2020706 |
AK009015.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2020706 |
BC060287.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
584230 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
76554 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
107206 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
592421 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
731170 |
AK133554.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
731170 |
AK136199.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
731170 |
AK145510.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
731170 |
AK162205.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
731170 |
NM_026480.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905753 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
810283 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
14646 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P111565 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P174709 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2020706 |
AK009015.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2020706 |
BC060287.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2007384 |
AF019564.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2007384 |
AK009015.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2007384 |
AK075915.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2007384 |
BC060287.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2007384 |
NM_001136080.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P335491 |
AF019564.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P335491 |
AK009015.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P335491 |
AK075915.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P335491 |
BC060287.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P335491 |
NM_001136080.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035754 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035760 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035804 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035883 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035880 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035806 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035759 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035805 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035881 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035763 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035807 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035802 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035797 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035756 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28035799 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07449204 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P174709 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P111565 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P335491 |
AF019564.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P335491 |
AK009015.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P335491 |
AK075915.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P335491 |
BC060287.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P335491 |
NM_001136080.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P115281 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P105649 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P111565 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P174709 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28035861 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28035860 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035862 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035752 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035812 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035875 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035756 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035781 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035865 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035872 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035793 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035844 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035818 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035738 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035821 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035866 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035764 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035842 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035745 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035763 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035834 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035876 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035780 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035881 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035823 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035807 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035861 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035797 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035850 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035817 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035815 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035830 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035826 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035860 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035805 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035796 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035831 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035828 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035778 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035708 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035767 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035883 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035854 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035858 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035754 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035832 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035880 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28035843 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09268939 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035815 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09140975 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035852 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09476252 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035869 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09392627 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035862 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035759 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035749 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035811 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035848 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035868 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035853 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035819 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035873 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09221446 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09221442 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09618524 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035870 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09392631 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09476248 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09140984 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035747 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09142859 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035809 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035837 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035804 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035860 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035763 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035854 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09285966 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035832 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035875 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09392630 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09530074 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09029983 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035872 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09092037 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035816 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09500137 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035876 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09476254 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035858 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035826 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09494191 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035741 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09476260 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035847 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035797 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035823 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035839 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035821 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09140978 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035760 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035846 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035834 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09268940 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09392625 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035754 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28035841 |
- |
- |