Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133667.2 | NP_598428.2 | 1 - 2 | 233 | 72 | |
GenBank mRNA | AF267660.1 | - | 1 - 2 | 194 | 72 | |
AK002647.1 | - | 1 - 2 | 178 | 72 | ||
BC021764.1 | - | 1 - 2 | 187 | 3488813 | 72 | |
CX223857.1 | - | 1 - 2 | 233 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |