Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133667.2 | NP_598428.2 | 3 - 4 | 451 | 59 | |
GenBank mRNA | AF267660.1 | - | 3 - 4 | 412 | 59 | |
AK002647.1 | - | 3 - 4 | 395 | 59 | ||
AK181188.1 | - | 2 - 3 | 195 | 59 | ||
AK192357.1 | - | 2 - 3 | 194 | 59 | ||
AK216417.1 | - | 2 - 3 | 194 | 59 | ||
AK218086.1 | - | 2 - 3 | 194 | 59 | ||
BC021764.1 | - | 3 - 4 | 405 | 3488813 | 59 | |
CX223857.1 | - | 3 - 4 | 451 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |