Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il13ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK080147.1 | - | 7 - 8 | 905 | 73 | |
AK089984.1 | - | 7 - 8 | 936 | 73 | ||
AK143292.1 | - | 7 - 8 | 905 | 73 | ||
AK154675.1 | - | 7 - 8 | 961 | 73 | ||
BC021472.1 | - | 3 - 4 | 359 | 73 | ||
BC052425.1 | - | 7 - 8 | 928 | 6835500 | 73 | |
BC059939.1 | - | 7 - 8 | 896 | 6477224 | 73 | |
S80963.1 | - | 7 - 8 | 933 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |