Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300008174.mm.1 |
NM_207205.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008174.mm.1 |
NM_207205.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008174.mm.1 |
TC0300001014.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008173.mm.1 |
NM_207205.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008173.mm.1 |
NM_207205.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008173.mm.1 |
TC0300001014.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008177.mm.1 |
NM_207205.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008177.mm.1 |
NM_207205.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008177.mm.1 |
TC0300001014.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008178.mm.1 |
NM_207205.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008178.mm.1 |
NM_207205.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008178.mm.1 |
TC0300001014.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008175.mm.1 |
NM_207205.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008175.mm.1 |
NM_207205.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008175.mm.1 |
TC0300001014.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008179.mm.1 |
NM_207205.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008179.mm.1 |
NM_207205.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008179.mm.1 |
TC0300001014.mm.1 |
MTA TC |