Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030150.2 | NP_084426.2 | 5 - 6 | 779 | 90 | |
GenBank mRNA | AF316999.1 | - | 4 - 5 | 678 | 90 | |
AK153342.1 | - | 5 - 6 | 708 | 90 | ||
BC029209.1 | - | 4 - 5 | 689 | 2651254 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |