Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172424.1 | NP_001165895.1 | 1 - 2 | 76 | 87 | |
GenBank mRNA | AA762067.1 | - | 1 - 2 | 149 | 87 | |
BC008980.1 | - | 1 - 2 | 76 | 3587135 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |