Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027349.3 | NP_081625.3 | 13 - 14 | 1721 | 196 | |
GenBank mRNA | AK142132.1 | - | 7 - 8 | 997 | 196 | |
AK162135.1 | - | 1819 | 196 | |||
BC158104.1 | - | 12 - 13 | 1601 | 9007388 | 196 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |