Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018854.4 | NP_061342.1 | 1 - 2 | 137 | 171 | |
GenBank mRNA | AK002397.1 | - | 1 - 2 | 138 | 171 | |
AK139942.1 | - | 1 - 2 | 174 | 171 | ||
BC002040.1 | - | 1 - 2 | 128 | 3590790 | 171 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |