Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033074.3 | NP_149065.2 | 4 - 5 | 568 | 81 | |
GenBank mRNA | AK010256.1 | - | 4 - 5 | 535 | 81 | |
AK011146.2 | - | 4 - 5 | 569 | 81 | ||
AK031064.1 | - | 4 - 5 | 532 | 81 | ||
AK152945.1 | - | 4 - 5 | 529 | 81 | ||
AK153314.1 | - | 4 - 5 | 540 | 81 | ||
AK166693.1 | - | 4 - 5 | 535 | 81 | ||
AK167597.1 | - | 4 - 5 | 577 | 81 | ||
AK168969.1 | - | 4 - 5 | 528 | 81 | ||
BC055371.1 | - | 4 - 5 | 554 | 30060749 | 81 | |
BC060176.1 | - | 507 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |