Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd40lg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011616.2 | NP_035746.2 | 2 - 3 | 300 | 102 | |
GenBank mRNA | BC119225.1 | - | 2 - 3 | 347 | 8733974 | 102 |
EU274582.1 | - | 2 - 3 | 288 | 102 | ||
X65453.2 | - | 2 - 3 | 300 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |