Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199045.1 | NP_001185974.1 | 8 - 9 | 1566 | 71 | |
NM_001199046.1 | NP_001185975.1 | 8 - 9 | 1460 | 71 | ||
NM_001199047.1 | NP_001185976.1 | 7 - 8 | 1473 | 71 | ||
NM_019756.3 | NP_062730.1 | 7 - 8 | 1367 | 71 | ||
GenBank mRNA | AB024329.1 | - | 6 - 7 | 1141 | 71 | |
AB031043.1 | - | 7 - 8 | 1172 | 71 | ||
AF081568.1 | - | 7 - 8 | 1207 | 71 | ||
AK016086.1 | - | 8 - 9 | 1261 | 71 | ||
AK030214.1 | - | 8 - 9 | 1566 | 71 | ||
AK169454.1 | - | 7 - 8 | 1368 | 71 | ||
AK170304.1 | - | 7 - 8 | 1332 | 71 | ||
AK171546.1 | - | 7 - 8 | 1334 | 71 | ||
BC026465.1 | - | 7 - 8 | 1350 | 4484099 | 71 | |
BC132325.1 | - | 7 - 8 | 1200 | 40130602 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |