Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500019124.mm.1 |
NM_001110216.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019124.mm.1 |
NM_001110216.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019124.mm.1 |
NM_007626.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019124.mm.1 |
NM_007626.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019124.mm.1 |
TC1500002309.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500019123.mm.1 |
NM_001110216.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019123.mm.1 |
NM_001110216.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019123.mm.1 |
NM_007626.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019123.mm.1 |
NM_007626.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019123.mm.1 |
TC1500002309.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500019121.mm.1 |
NM_001110216.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019121.mm.1 |
NM_001110216.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019121.mm.1 |
TC1500002309.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500019118.mm.1 |
NM_001076789.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019118.mm.1 |
NM_001076789.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019118.mm.1 |
NM_001110216.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019118.mm.1 |
NM_001110216.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019118.mm.1 |
NM_007626.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019118.mm.1 |
NM_007626.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019118.mm.1 |
TC1500002309.mm.1 |
MTA TC |