Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080118.1 | NP_001073587.1 | 2 - 3 | 341 | 71 | |
NM_013634.2 | NP_038662.2 | 2 - 3 | 206 | 71 | ||
NM_134027.2 | NP_598788.2 | 2 - 3 | 341 | 71 | ||
GenBank mRNA | AF000294.1 | - | 2 - 3 | 206 | 71 | |
AF332073.1 | - | 1 - 2 | 86 | 71 | ||
AF332074.1 | - | 1 - 2 | 86 | 71 | ||
AK005207.1 | - | 2 - 3 | 325 | 71 | ||
AK049203.1 | - | 2 - 3 | 332 | 71 | ||
AK054437.1 | - | 2 - 3 | 342 | 71 | ||
AK147199.1 | - | 2 - 3 | 318 | 71 | ||
BB846904.1 | - | 2 - 3 | 256 | 71 | ||
BC021440.1 | - | 2 - 3 | 312 | 71 | ||
BC079636.1 | - | 2 - 3 | 311 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |