Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080118.1 | NP_001073587.1 | 6 - 7 | 636 | 138 | |
NM_013634.2 | NP_038662.2 | 6 - 7 | 501 | 138 | ||
NM_134027.2 | NP_598788.2 | 6 - 7 | 636 | 138 | ||
GenBank mRNA | AF000294.1 | - | 6 - 7 | 501 | 138 | |
AF332073.1 | - | 5 - 6 | 381 | 138 | ||
AF332074.1 | - | 5 - 6 | 381 | 138 | ||
AK005207.1 | - | 6 - 7 | 137 | |||
AK049203.1 | - | 6 - 7 | 627 | 138 | ||
AK054437.1 | - | 6 - 7 | 637 | 138 | ||
AK147199.1 | - | 6 - 7 | 613 | 138 | ||
BC079636.1 | - | 6 - 7 | 606 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |