Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159319.2 | NP_001152791.1 | 6 - 7 | 828 | 91 | |
NM_145121.3 | NP_660099.2 | 6 - 7 | 828 | 91 | ||
GenBank mRNA | AY094172.1 | - | 6 - 7 | 626 | 91 | |
AY094173.1 | - | 6 - 7 | 626 | 91 | ||
CA316737.1 | - | 5 - 6 | 522 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mus musculus Probe spans exons |