Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081030.2 | NP_001074499.2 | 9 - 10 | 1213 | 63 | |
NM_001170561.1 | NP_001164032.1 | 9 - 10 | 1213 | 63 | ||
GenBank mRNA | AK129485.1 | - | 9 - 10 | 1157 | 63 | |
AK147332.1 | - | 9 - 10 | 1174 | 63 | ||
BC157935.1 | - | 9 - 10 | 1103 | 9088115 | 63 | |
BC158013.1 | - | 9 - 10 | 1103 | 9007320 | 63 | |
BC172094.1 | - | 9 - 10 | 1103 | 9054380 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |