Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144851.5 | NP_659100.1 | 13 - 14 | 1730 | 85 | |
GenBank mRNA | AK028565.1 | - | 13 - 14 | 1472 | 85 | |
AK028762.1 | - | 5 - 6 | 2704 | 85 | ||
AK035723.1 | - | 14 - 15 | 1913 | 85 | ||
AK054521.1 | - | 14 - 15 | 1862 | 85 | ||
BC023129.1 | - | 12 - 13 | 1444 | 3595534 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |