Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048178.2 | NP_001041643.1 | 6 - 7 | 828 | 99 | |
NM_001286370.1 | NP_001273299.1 | 6 - 7 | 529 | 99 | ||
NM_010284.3 | NP_034414.2 | 6 - 7 | 828 | 99 | ||
GenBank mRNA | AK143886.1 | - | 6 - 7 | 530 | 99 | |
AK143925.1 | - | 6 - 7 | 626 | 99 | ||
BC024375.1 | - | 6 - 7 | 663 | 99 | ||
BC056645.1 | - | 6 - 7 | 644 | 99 | ||
BC075720.1 | - | 6 - 7 | 676 | 5044060 | 99 | |
M31680.1 | - | 6 - 7 | 564 | 99 | ||
M33324.1 | - | 6 - 7 | 564 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |