Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175251.4 | NP_780460.3 | 2 - 3 | 356 | 91 | |
GenBank mRNA | AK007269.1 | - | 2 - 3 | 358 | 91 | |
BU056807.1 | - | 2 - 3 | 163 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |